Learn More
Amsbio LLC StemFit Media For Mesenchymal Stem Cells (MSC) (AOF, chemically defined medium for stem cell culture)
Catalog No. 502041691
Encompass
StemFit For Mesenchymal Stem Cells (MSC) is a chemically defined & animal origin free medium optimized for the expansion & maintenance of human bone marrow-derived MSC (BM-MSC), umbilical cord-derived MSC (UC-MSC), & adipose-derived stem cells (ADSC), under serum-free, human platelet lysate free conditions. StemFit For MSC offers exceptional cell expansion with high levels of marker expression & differentiation potential. For best results, we recommend using StemFit For MSC with iMatrix-511 as a cell culture substrate. MAIN FEATURES: The optimal medium for expansion and maintenance of mesenchymal stem cells (MSCs) | Superior cell expansion compared to serum-containing media | Chemically defined medium minimizes lot-to-lot variation | Suitable for use with umbilical cord-derived MSCs. human bone marrow-derived MSCs, and adipose-derived stem cells | Low risk of immunogenic contamination due to animal origin-free formula.
Catalog No. 50-204-1691 Supplier Amsbio LLC Supplier No. SFMSCA3
StemFit For Mesenchymal Stem Cells (MSC) is a chemically defined & animal origin free medium optimized for the expansion & maintenance of human bone marrow-derived MSC (BM-MSC), umbilical cord-derived MSC (UC-MSC), & adipose-derived stem cells (ADSC), under serum-free, human platelet lysate free conditions. StemFit For MSC offers exceptional cell expansion with high levels of marker expression & differentiation potential. For best results, we recommend using StemFit For MSC with iMatrix-511 as a cell culture substrate. MAIN FEATURES: The optimal medium for expansion and maintenance of mesenchymal stem cells (MSCs) | Superior cell expansion compared to serum-containing media | Chemically defined medium minimizes lot-to-lot variation | Suitable for use with umbilical cord-derived MSCs. human bone marrow-derived MSCs, and adipose-derived stem cells | Low risk of immunogenic contamination due to animal origin-free formula.