Learn More
S BIO STEMFULL™ 15mL Centrifuge Tube
Catalog No. NC1320014
Encompass
STEMFULL™ is an ultra-hydrophilic polymer-coated centrifuge tube designed to minimize nonspecific cell adhesion to tube walls and thus ensure high cell recovery after centrifugation. The ultra-hydrophilic polymer is covalently bonded to the tube surface, reducing the risk of coating material leaching. STEMFULL™ has been proven effective for recovering strongly adherent cells such as human stem cells, thereby improving recovery efficiency and reproducibility in stem cell culture and processing.
Catalog No. NC1320014 Supplier S BIO Supplier No. MS90150
STEMFULL™ is an ultra-hydrophilic polymer-coated centrifuge tube designed to minimize nonspecific cell adhesion to tube walls and thus ensure high cell recovery after centrifugation. The ultra-hydrophilic polymer is covalently bonded to the tube surface, reducing the risk of coating material leaching. STEMFULL™ has been proven effective for recovering strongly adherent cells such as human stem cells, thereby improving recovery efficiency and reproducibility in stem cell culture and processing.