Learn More
Reprocell STEMOLECULE PD0325901 2 MG
Catalog No. NC9753132
Encompass
PD0325901 is a small molecule targeting mitogen-activated protein kinase (MaPK/ERK kinase or MEK) with potential antineoplastic activity. PD0325901, a derivative of MEK inhibitor CI-1040, selectively binds to and inhibits MEK, which may result in the inhibition of the phosphorylation and activation of MaPK/ERK and the inhibition of tumor cell proliferation1,2. along with the aLK5 inhibitor SB431542, PD0325901 has also been shown to increase the efficiency of reprogramming human primary fibroblasts into induced pluripotent stem (iPS) cells3. 2mg.
Catalog No. NC9753132 Supplier Reprocell Supplier No. 040006
PD0325901 is a small molecule targeting mitogen-activated protein kinase (MaPK/ERK kinase or MEK) with potential antineoplastic activity. PD0325901, a derivative of MEK inhibitor CI-1040, selectively binds to and inhibits MEK, which may result in the inhibition of the phosphorylation and activation of MaPK/ERK and the inhibition of tumor cell proliferation1,2. along with the aLK5 inhibitor SB431542, PD0325901 has also been shown to increase the efficiency of reprogramming human primary fibroblasts into induced pluripotent stem (iPS) cells3. 2mg.