Learn More
Reprocell STEMOLECULE SB431542
Catalog No. NC9993293
Encompass
SB431542 is an inhibitor of the transforming growth factor-beta 1 (TGF-β1) activin receptor-like kinases (ALKs). It is a selective and potent inhibitor of ALK-4, -5 and -7. SB431542 inhibits endogenous activin and TGF-β signaling without affecting more divergent BMP signaling utilizing ALK-1, -2, -3, and -61,2. SB431542 stimulates proliferation, differentiation, and sheet formation of endothelial cells derived from embryonic stem cells3.
Catalog No. NC9993293 Supplier Reprocell Supplier No. 040010
SB431542 is an inhibitor of the transforming growth factor-beta 1 (TGF-β1) activin receptor-like kinases (ALKs). It is a selective and potent inhibitor of ALK-4, -5 and -7. SB431542 inhibits endogenous activin and TGF-β signaling without affecting more divergent BMP signaling utilizing ALK-1, -2, -3, and -61,2. SB431542 stimulates proliferation, differentiation, and sheet formation of endothelial cells derived from embryonic stem cells3.