Learn More
Reprocell STEMOLECULE WNT INHB IWP-2 2MG
Catalog No. NC9086357
Encompass
Stemolecule™ Wnt Inhibitor IWP-2. Stemolecule Wnt Inhibitor IWP-2 was identified in a high throughput screen for antagonists of the Wnt/β-catenin pathway. Wnt Inhibitor IWP-2 prevents palmitylation of Wnt proteins by Porcupine (Porcn), a membrane-bound O-acyltransferase, thereby blocking Wnt secretion and activity. It also blocks phosphorylation of the Lrp6 receptor and accumulation of both Dvl2 and β-catenin1.
Catalog No. NC9086357 Supplier Reprocell Supplier No. 040034
Stemolecule™ Wnt Inhibitor IWP-2. Stemolecule Wnt Inhibitor IWP-2 was identified in a high throughput screen for antagonists of the Wnt/β-catenin pathway. Wnt Inhibitor IWP-2 prevents palmitylation of Wnt proteins by Porcupine (Porcn), a membrane-bound O-acyltransferase, thereby blocking Wnt secretion and activity. It also blocks phosphorylation of the Lrp6 receptor and accumulation of both Dvl2 and β-catenin1.