Learn More
Reprocell Stemolecule XAV939, 284028-89-3, MFCD16879017, 2 mg
Catalog No. NC9086361
Encompass
Formula: C14H11F3N2OS, MW: 312.31, White powder, Names: 2-(4-(trifluoromethyl)phenyl)-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol.
Catalog No. NC9086361 Supplier Reprocell Supplier No. 040046
Formula: C14H11F3N2OS, MW: 312.31, White powder, Names: 2-(4-(trifluoromethyl)phenyl)-7,8-dihydro-5H-thiopyrano[4,3-d]pyrimidin-4-ol.