Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ StemPro™-34 SFM (1X)

Catalog No. 10639011
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-639-011 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-639-011 Supplier Gibco™ Supplier No. 10639011
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A serum-free medium specifically formulated to support the development of human hematopoietic cells in culture.

  • A serum-free medium specifically formulated to support the development of human hematopoietic cells in culture
  • This includes HSC and progenitor cells isolated from bone marrow, neonatal cord blood, and peripheral blood
  • StemPro-34 SFM is a flexible cell culture media that can be used for a wide variety of applications
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hematopoietic Stem Cell, Hematopoietic Cell, Bone Marrow
Classification Serum-free
Form Liquid
Product Type Stem Cell Serum Free Media (SFM)
Serum Level Serum-free
With Additives Phenol Red
Without Additives No Glutamine
Product Line StemPro
Quantity 500 mL
Endotoxin Level ≤ 10 EU/mL
Species Human
Content And Storage • 500 ml StemPro™-34 Serum Free Medium (store at 2°C to 8°C in the dark)
• 13 ml StemPro™-34 Nutrient Supplement (store at -5°C to -20°C in the dark)

Store the complete medium at 2°C to 8°C in the dark.
Culture Type Stationary Suspension Culture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.