Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ StemPro™ EZPassage™ Disposable Stem Cell Passaging Tool

Catalog No. 23181010
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-181-010 10 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-181-010 Supplier Gibco™ Supplier No. 23181010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Novel human embryonic stem cell passaging device that makes manual passaging of stem cell colonies more rapid and reproducible.

  • Made of a proprietary, cell culture-safe, and inert material that enables cutting of embryonic stem cell colonies growing on feeder cells or other substrates into uniform pieces for optimal passaging.
  • Outperforms existing manual and enzymatic passing methods in speed, uniformity of passaged colonies, and reliability.
  • Manually passage stem cell colonies in a fraction of the time compared with standard techniques.
  • Cut stem cell colonies into pieces of uniform size, making passaging of stem cells more reproducible.
  • Control the size for better passaging or to make embryoid bodies.
  • Cell culture-safe Passaging Tools are ready for use and packaged individually in gamma-irradiated sleeves.
  • One-time use.
  • Do not sterilize (alcohol or autoclave)
TRUSTED_SUSTAINABILITY

Specifications

Material Proprietary, Cell Culture-Safe, Inert Material
Sterility Sterile
For Use With (Application) Manual Passaging Stem Cell Colonies
Disposable Single-use
Product Type Stem Cell Passaging Tool
Cell Type Stem Cell
Product Line EZPassage, StemPro
Quantity 10 units
In the StemPro EZPassage Disposable Stem Cell Passaging Tool (Cat. No. 23181010), what is the exact distance between the blades of the roller?

The tool has ˜230 micrometers between each blade. We don't recommend using the tool on anything smaller than a 6-well plate. As far as the thickness at the base of each blade relative to the distance between the blade's edges, we don't have data on that.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.