Learn More
STEMCELL Technologies StemSpan™ Erythroid Expansion Supplement (100X), Size: 1 mL
SDP
Supplier:  STEMCELL Technologies 02692
Encompass
StemSpan™ Erythroid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of erythroid progenitor cells from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.When added to serum-free medium, StemSpan™ Erythroid Expansion Supplement typically promotes the production of thousands of erythroid cells per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.StemSpan™ Erythroid Expansion Supplement (100X) is intended for use in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-ACF Erythroid Expansion Medium (Catalog #09860)
Catalog No. NC1568103