Learn More
Cayman Chemical StemRegenin 1 Other Receptor M
SDP
Supplier:  Cayman Chemical 1062510 MG
Encompass_Preferred
A purine derivative that reversibly antagonizes aryl hydrocarbon receptor signaling (IC50 = 127 nM in CD34+ cells); induces a 50-fold increase in human embryonic stem cells expressing CD34 (EC50 = 120 nM) and a 17-fold increase in stem cells that retain the ability to engraft immunodeficient mice
Catalog No. 50-289-0212