Learn More
Apexbio Technology LLC StemRegenin 1 (SR1) 1227633-49-9 200mg
SDP
Catalog No. 5000061689
Encompass_Preferred
StemRegenin 1 (SR1 CAS 1227633-49-9) is a purine derivative that functions as an antagonist of the aryl hydrocarbon receptor (AhR) SR1 inhibits AhR signaling in CD34 hematopoietic progenitor cells with an IC50 value of approximately 127 nM Studies demonstrate that SR1 promotes the ex vivo expansion of CD34 cells significantly increasing both the number of CD34-expressing hematopoietic stem cells (HSCs) and their engraftment potential in immunodeficient mouse models Additionally SR1 enables differentiation and robust production of multiple dendritic cell subsets including plasmacytoid dendritic cells (pDCs) and BDCA1 /BDCA3 myeloid DCs from CD34 progenitors indicating its utility for dendritic cell research and hematopoietic stem cell biology
Catalog No. 50-000-61689 Supplier Apexbio Technology LLC Supplier No. A8224200
StemRegenin 1 (SR1 CAS 1227633-49-9) is a purine derivative that functions as an antagonist of the aryl hydrocarbon receptor (AhR) SR1 inhibits AhR signaling in CD34 hematopoietic progenitor cells with an IC50 value of approximately 127 nM Studies demonstrate that SR1 promotes the ex vivo expansion of CD34 cells significantly increasing both the number of CD34-expressing hematopoietic stem cells (HSCs) and their engraftment potential in immunodeficient mouse models Additionally SR1 enables differentiation and robust production of multiple dendritic cell subsets including plasmacytoid dendritic cells (pDCs) and BDCA1 /BDCA3 myeloid DCs from CD34 progenitors indicating its utility for dendritic cell research and hematopoietic stem cell biology