Learn More
Reprocell StemRNA Human iPSC 771-3G (1 vial)
Catalog No. NC3450519
Encompass
Ready to use IPS Cells made using the Cutting-Edge StemRNA 3rd Gen Reprogramming Technology. Taking full advantage of the power of the 3rd Gen Technology, these cells feature:No possibility of integration or retention of Reprogramming VectorsImmediate thawing into NutriStem hPSC XF Culture medium (Cat. No. 01-0005) in iMatrix-511 substrate (Cat. No. NP892-011) with easy conversion to other standard iPSC Media.The possibility of saving significant time compared to reprogramming your own cells.
Catalog No. NC3450519 Supplier Reprocell Supplier No. RCRP005N
Add to Cart
Add to Cart
Ready to use IPS Cells made using the Cutting-Edge StemRNA 3rd Gen Reprogramming Technology. Taking full advantage of the power of the 3rd Gen Technology, these cells feature:No possibility of integration or retention of Reprogramming VectorsImmediate thawing into NutriStem hPSC XF Culture medium (Cat. No. 01-0005) in iMatrix-511 substrate (Cat. No. NP892-011) with easy conversion to other standard iPSC Media.The possibility of saving significant time compared to reprogramming your own cells.