Learn More
STEMCELL Technologies STEMscript cDNA Synthesis Kit with Random Primers, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 79004
Encompass
Use STEMscript cDNA Synthesis Kit with Random Primers for reverse transcription of RNA without poly(A) tails (e.g. rRNA tRNA degraded RNA RNA isolated from prokaryotic sources and RNA with known secondary structures) prior to qPCR or RNA sequencing.
Catalog No. 50-239-0219