Learn More
STEMCELL Technologies StemSpan™ CC100, Size: 1 mL
SDP
Catalog No. NC9421569
Encompass
StemSpan™ CC100 cytokine cocktail contains a combination of both early- and late-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate.When added to serum-free medium, StemSpan™ CC100 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow, and is recommended if high cell yields and large numbers of CD34+ cells are desired.We recommend using StemSpan™ CC100 in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-AOF (Catalog #100-0130)
Catalog No. NC9421569 Supplier STEMCELL Technologies Supplier No. 02690
StemSpan™ CC100 cytokine cocktail contains a combination of both early- and late-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate.When added to serum-free medium, StemSpan™ CC100 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow, and is recommended if high cell yields and large numbers of CD34+ cells are desired.We recommend using StemSpan™ CC100 in combination with any of the following StemSpan™ media:• StemSpan™ SFEM (Catalog #09600)• StemSpan™ SFEM II (Catalog #09605)• StemSpan™-XF (Catalog #100-0073)• StemSpan™-AOF (Catalog #100-0130)