Learn More
STEMCELL Technologies StemSpan™ HSC Plus Supplement, Animal origin-free supplement for the expansion of human hematopoietic stem and progenitor cells
SDP
Catalog No. NC3807134
Encompass
Enhance your hematopoietic stem and progenitor cell (HSPC) research workflow with StemSpan™ HSC Plus Supplement—a defined mix of small molecules formulated to improve the yield of hematopoietic stem cells (HSCs) when combined with a cytokine-containing medium. This formulation is optimized to support various workflows, including those involving gene editing, engraftment studies, or expansion to increase cell yields. When used with a cytokine-containing medium in ex vivo culture, StemSpan™ HSC Plus Supplement promotes the expansion of HSCs while preserving their functional potential, helping you achieve higher yields and consistent results.
Catalog No. NC3807134 Supplier STEMCELL Technologies Supplier No. 1001694
Add to Cart
Add to Cart
Enhance your hematopoietic stem and progenitor cell (HSPC) research workflow with StemSpan™ HSC Plus Supplement—a defined mix of small molecules formulated to improve the yield of hematopoietic stem cells (HSCs) when combined with a cytokine-containing medium. This formulation is optimized to support various workflows, including those involving gene editing, engraftment studies, or expansion to increase cell yields. When used with a cytokine-containing medium in ex vivo culture, StemSpan™ HSC Plus Supplement promotes the expansion of HSCs while preserving their functional potential, helping you achieve higher yields and consistent results.