Learn More
STEMCELL Technologies StemSpan™ Leukemic Cell Culture Kit, Size: 1 Kit
SDP
Catalog No. 501974450
Encompass
StemSpan™ Leukemic Cell Culture Kit has been developed for the in vitro culture and expansion of malignant cells and has been tested on chronic myeloid leukemia (CML) and acute myeloid leukemia (AML) samples. This optimized protocol allows users to expand, culture, and use malignant cells for drug screening. StemSpan™ Leukemic Cell Culture Kit includes the serum-free medium SFEM II (Catalog #09605), StemSpan™ CD34+ Expansion Supplement (10X; Catalog #02691), and small molecule UM729 (Catalog #72332).
Catalog No. 50-197-4450 Supplier STEMCELL Technologies Supplier No. 09720
StemSpan™ Leukemic Cell Culture Kit has been developed for the in vitro culture and expansion of malignant cells and has been tested on chronic myeloid leukemia (CML) and acute myeloid leukemia (AML) samples. This optimized protocol allows users to expand, culture, and use malignant cells for drug screening. StemSpan™ Leukemic Cell Culture Kit includes the serum-free medium SFEM II (Catalog #09605), StemSpan™ CD34+ Expansion Supplement (10X; Catalog #02691), and small molecule UM729 (Catalog #72332).