Learn More
STEMCELL Technologies StemSpan Myeloid Expansion Supplement (100X), Size: 1 mL
SDP
Supplier:  STEMCELL Technologies 02693
Encompass
StemSpan Myeloid Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human myeloid progenitor cells generating large numbers of CD15 granulocytes from CD34 cells isolated from human cord blood (CB) or bone marrow (BM) samples.
Catalog No. 50-239-0338