Learn More
FUJIFILM Biosciences StemSure Mono Solution X100
Catalog No. 5000450367
Encompass_Preferred
Monothioglycerol MTG can be used as an alternative for 2-mercaptoethanol 2ME since it exhibits comparable performance to 2ME when it is added to a medium in the culture of mouse ES D3 strain cells This product is added to the culture medium as a reducing agent
Catalog No. 50-004-50367 Supplier FUJIFILM Biosciences Supplier No. 19515791
Monothioglycerol MTG can be used as an alternative for 2-mercaptoethanol 2ME since it exhibits comparable performance to 2ME when it is added to a medium in the culture of mouse ES D3 strain cells This product is added to the culture medium as a reducing agent