Learn More
Worthington Biochemical Corporation STEMXYME 2 COLLAGENASE 50MG
SDP
Catalog No. 50592430
Encompass
A specialized combination of Animal Origin-Free Clostridium histolyticum collagenase (Code: CLSAFB) and animal origin free Bacillus polymyxa neutral protease with a minimum of 250 CLS units and 2,000 caseinase units per mg dry weight. Designed for stem cell and other primary cell isolations and bioprocessing applications where introduction of potential animal derived pathogens must be prevented. 2593-92-3
Catalog No. 50-592-430 Supplier Worthington Biochemical Corporation Supplier No. LS004112
A specialized combination of Animal Origin-Free Clostridium histolyticum collagenase (Code: CLSAFB) and animal origin free Bacillus polymyxa neutral protease with a minimum of 250 CLS units and 2,000 caseinase units per mg dry weight. Designed for stem cell and other primary cell isolations and bioprocessing applications where introduction of potential animal derived pathogens must be prevented. 2593-92-3