Learn More
Selleck Chemical LLC Stenabolic (SR9009) 5mg 1379686-30-2
SDP
Supplier:  Selleck Chemical LLC S86925MG
Encompass_Preferred
Stenabolic (SR9009) is a synthetic REV-ERB agonist with IC50s of 670 nM and 800 nM for REV-ERB-α and for REV-ERB-β, respectively.
Catalog No. 50-194-8221