Learn More
ATCC Stenotrophomonas; maltophilia; 6 x 0.3mL
Catalog No. 502383850
Encompass
ATCC 17666-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 17666™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-3850 Supplier ATCC Supplier No. 17666MINIPACK
ATCC 17666-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 17666™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.