Learn More
ATCC Stenotrophomonas maltophilia; Strain: 8402014
Catalog No. 502382204
Encompass
Quality control strain for bioMerieux API 20E kits.
Catalog No. 50-238-2204 Supplier ATCC Supplier No. 51331
Quality control strain for bioMerieux API 20E kits.