Learn More
ATCC Stenotrophomonas; maltophilia; Strain 300
Catalog No. 502382533
Encompass
Stenotrophomonas maltophilia strain 300 [X-56] is a whole-genome sequenced bacterial strain that was isolated from a contaminated tissue culture. This strain produces nitrate reductase and has applications as a quality control strain.
Catalog No. 50-238-2533 Supplier ATCC Supplier No. 17666
Stenotrophomonas maltophilia strain 300 [X-56] is a whole-genome sequenced bacterial strain that was isolated from a contaminated tissue culture. This strain produces nitrate reductase and has applications as a quality control strain.