Learn More
Grainger Step Bushing 1/2 Dia Black Rubber PK50
Catalog No. 502767776
Encompass_Preferred
The Step Bushing Door Mounted is a door-mounted step bushing that provides a smooth, quiet operation for doors. It is made of thermoplastic and has a low-friction internal liner. The step bushing has a 1-3/8" ID and a 2-1/4" OD. It is designed for use with 1-3/8" shafts on commercial and industrial doors. This product helps reduce noise and friction for door pivots. It can support door weights up to 400 lbs. The step bushing is durable, low-maintenance, and ensures smooth door operation. This product is ideal for warehouses, manufacturing facilities, or other commercial/industrial buildings using mounted doors.
Catalog No. 50-276-7776 Supplier Grainger Supplier No. 16ZD70
The Step Bushing Door Mounted is a door-mounted step bushing that provides a smooth, quiet operation for doors. It is made of thermoplastic and has a low-friction internal liner. The step bushing has a 1-3/8" ID and a 2-1/4" OD. It is designed for use with 1-3/8" shafts on commercial and industrial doors. This product helps reduce noise and friction for door pivots. It can support door weights up to 400 lbs. The step bushing is durable, low-maintenance, and ensures smooth door operation. This product is ideal for warehouses, manufacturing facilities, or other commercial/industrial buildings using mounted doors.