Learn More
Grainger Milwaukee Tool Step Drill Bit Set, Dual-Flute, Black Oxide, 1/8 in. to 1-1/8 in., 6/Pk.
Catalog No. 502783073
Encompass_Preferred
This 6-piece step drill bit set is designed for drilling small and large-diameter holes in steel and plastic. Featuring a jam-free dual-flute design, these bits deliver faster hole creation and increased control, optimized for use with cordless drills at high speed. The bits are finished in black oxide for enhanced durability, reduced heat generation, and longer bit life.
  • Dual-flute design provides up to 2X faster drilling and up to 4X longer bit life.
  • Rapid strike tip ensures fast, accurate starts with minimal heat generation.
  • Laser-engraved reference marks allow for precise hole sizing.
  • Three-flat secure-grip shanks reduce bit slippage in the drill chuck.
  • Black oxide coating enhances durability and improves hole quality.
  • Includes six step drill bits and a carrying case.
Catalog No. 50-278-3073 Supplier Grainger Supplier No. 19RK85
This 6-piece step drill bit set is designed for drilling small and large-diameter holes in steel and plastic. Featuring a jam-free dual-flute design, these bits deliver faster hole creation and increased control, optimized for use with cordless drills at high speed. The bits are finished in black oxide for enhanced durability, reduced heat generation, and longer bit life.
  • Dual-flute design provides up to 2X faster drilling and up to 4X longer bit life.
  • Rapid strike tip ensures fast, accurate starts with minimal heat generation.
  • Laser-engraved reference marks allow for precise hole sizing.
  • Three-flat secure-grip shanks reduce bit slippage in the drill chuck.
  • Black oxide coating enhances durability and improves hole quality.
  • Includes six step drill bits and a carrying case.