Learn More
LabScientific Step Up Slide Warmer 120V; 110V; (USA and Canada); Surface holds about 40 slides. Heats up to 70 C.
Catalog No. NC1043525
Encompass
Step Up Slide Warmer 120V; 110V; (USA and Canada); Surface holds about 40 slides. Heats up to 70 C. Clean, dry and ventilated. Stable work surface. Relative humidity:≤80%. Setting time range 1-240min. Dimensions 14 x 14 x 4 in. (35 x 35 x 10cm.). Ship weight 22 pounds.
Catalog No. NC1043525 Supplier LabScientific Supplier No. XH2003
Step Up Slide Warmer 120V; 110V; (USA and Canada); Surface holds about 40 slides. Heats up to 70 C. Clean, dry and ventilated. Stable work surface. Relative humidity:≤80%. Setting time range 1-240min. Dimensions 14 x 14 x 4 in. (35 x 35 x 10cm.). Ship weight 22 pounds.