Learn More
Med Vet International Step Up Slide Warmer
Catalog No. 503046442
Encompass_Preferred
Our Step Up™ Slide Warmer’s unique patent pending design helps you save space while still meeting a busy lab’s needs.The Step Up™ holds twice as many slides as a flat unit of the same size and also makes retrieving your slides easier since the end of the slide is not on the flat surface.
Catalog No. 50-304-6442 Supplier Med Vet International Supplier No. XH2003
Our Step Up™ Slide Warmer’s unique patent pending design helps you save space while still meeting a busy lab’s needs.The Step Up™ holds twice as many slides as a flat unit of the same size and also makes retrieving your slides easier since the end of the slide is not on the flat surface.