Learn More
Cayman Chemical StephacIdIn B 2.5mg
SDP
Catalog No. 502988062
Encompass_Preferred
A fungal metabolite; converted to avrainvillamide in vitro; cytotoxic to a variety of cancer cells, including testosterone-independent PC3 and -sensitive LNCaP prostate cancer cells (IC50s = 0.37 and 0.06 µM, respectively) and estradiol-independent SK-BR-3 and -sensitive MCF-7 breast cancer cells (IC50s = 0.32 and 0.27 µM, respectively); induces apoptosis in HepG2 and Huh7 cells at 4 µM
Catalog No. 50-298-8062 Supplier Cayman Chemical Supplier No. 316562.5 MG
A fungal metabolite; converted to avrainvillamide in vitro; cytotoxic to a variety of cancer cells, including testosterone-independent PC3 and -sensitive LNCaP prostate cancer cells (IC50s = 0.37 and 0.06 µM, respectively) and estradiol-independent SK-BR-3 and -sensitive MCF-7 breast cancer cells (IC50s = 0.32 and 0.27 µM, respectively); induces apoptosis in HepG2 and Huh7 cells at 4 µM