Learn More
Research Products International Corp Stepped Tubing Connectors for 1/4 Inch to 1/2 Inch Tubing, Polypropylene, 12 per Bag
SDP
Supplier:  Research Products International Corp F19562
Encompass
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.). Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.
Catalog No. 50-998-414