Learn More
Research Products International Corp Stepped Tubing Connectors, 3/16x3/8, 12/pk
SDP
Catalog No. 501978328
Encompass
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.). Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.
Catalog No. 50-197-8328 Supplier Research Products International Corp Supplier No. F19559
These fittings ensure a tight, secure connection between tubing having different inner diameters (I.D.). Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.