Learn More
Waters Corp Waters Ion Guide Tools, Stepwave, Removal & Insertion Kit
Catalog No. 50466661 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides essential tools for the maintenance of the Stepwave ion guide. It includes a seal breaker/locator tool and an ion guide removal/insertion guide to facilitate proper servicing.
  • Includes a seal breaker/locator tool
  • Includes an ion guide removal/insertion guide
Catalog No. 50-466-661 Supplier Waters Corp Supplier No. 700005596
This kit provides essential tools for the maintenance of the Stepwave ion guide. It includes a seal breaker/locator tool and an ion guide removal/insertion guide to facilitate proper servicing.
  • Includes a seal breaker/locator tool
  • Includes an ion guide removal/insertion guide