Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI STER-AHOL Disinfectant

Catalog No. 19546858 Shop All VAI Products
Change view
Click to view available options
Volume (English):
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-546-858 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-546-858 Supplier VAI Supplier No. DSTERWFI705G
  • $794.00
    $635.65 / Each of 1

    Save $158.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

STER-AHOL Disinfectant is formulated to 70% Denatured Ethanol and 30% USP Water for Injection. Can be used for the disinfection and decontamination of cleanrooms, controlled areas, hard non-porous environmental surfaces, and many other applications that require the use of a sterile alcohol solution.

  • Effective sanitizer on inanimate, non-porous, non-food contact surfaces
  • Sterile pharmaceutical cleanroom formula
  • Phosphate free

Specifications

Sterility Sterile
Composition Denatured Ethanol
Concentration 70%
Filtered Filtered
Volume (English) 5 gal.
Volume (Metric) 19 L
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.