Learn More
Cole-Parmer Sterilized Air Filter; 0.2 um Pore, 50 mm Dia, 10/pk
Supplier:  Cole-Parmer 1594522
Encompass_Preferred
Sterile filter with hydrophobic PTFE membrane for degassing bioreactors. Stepped hose barb connections accept 1/4" to 3/8" inner diameter tubing.
Catalog No. NC1987173