Learn More
Hanna Pharmaceutical Supply Co., Inc. CROSSTEX DUO-CHECK STERILIZATION POUCHES
Catalog No. NC0304165
Encompass_Preferred
7.5"x13"/19x33 cm Duo-Check™ Self-Seal Sterilization Pouches Built-in internal/external process indicators that change color when exposed to a specific temperatureManufactured from medical grade paper with extra-wide chevron to protect against instrument breaching, thumb-notch for easy opening, 6 corner tack seals to prevent curling and other potential contaminants from entering pouch upon opening and a perforated fold to help ensure the adhesive strip forms an airtight sealOnce sterilized, contents will remain sterile for up to 3 years provided the integrity of the pouch has not been compromisedFor use with steam and EtO sterilization processes only 200/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC0304165 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. HFGSCL
7.5"x13"/19x33 cm Duo-Check™ Self-Seal Sterilization Pouches Built-in internal/external process indicators that change color when exposed to a specific temperatureManufactured from medical grade paper with extra-wide chevron to protect against instrument breaching, thumb-notch for easy opening, 6 corner tack seals to prevent curling and other potential contaminants from entering pouch upon opening and a perforated fold to help ensure the adhesive strip forms an airtight sealOnce sterilized, contents will remain sterile for up to 3 years provided the integrity of the pouch has not been compromisedFor use with steam and EtO sterilization processes only 200/BX
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.