Learn More
USA Scientific Inc 1.5 mL conical screw cap tube, natural, sterile
Catalog No. NC9656954
Encompass
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural with natural cap, sterile, 10 bags of 50 (500 tubes and caps)
Catalog No. NC9656954 Supplier USA Scientific Inc Supplier No. 14158710
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural with natural cap, sterile, 10 bags of 50 (500 tubes and caps)