Learn More
ENVIRO MIST INC STERAMIST BIT SOL 4X1GAL/CS
Catalog No. NC2330789
Encompass
SteraMist BIT Solution Cartridge 1GAL, 4 cartridges per CS. Steramist provides everything that is important for surface disinfection.High efficacy. Delivers six log kill 99.9999% reduction of pathogens.No residue left behind, ABSOLUTELY NONE. Aerates and evaporates leaving Surfaces free of slimy, sticky,tacky or visible residue. No rinse or wipe needed.
Catalog No. NC2330789 Supplier ENVIRO MIST INC Supplier No. 987598
Add to Cart
Add to Cart
SteraMist BIT Solution Cartridge 1GAL, 4 cartridges per CS. Steramist provides everything that is important for surface disinfection.High efficacy. Delivers six log kill 99.9999% reduction of pathogens.No residue left behind, ABSOLUTELY NONE. Aerates and evaporates leaving Surfaces free of slimy, sticky,tacky or visible residue. No rinse or wipe needed.