Learn More
Accuflow Systems STERAMIST BIT-400 ready-to-use solution, 1 Gallon, 4 bottles/case
Catalog No. NC1442001
Encompass_Preferred
The BIT-400 a ready-to-use solution, is to be used strictly in conjuction with SteraMist™ disinfection/decontamination system. It is a Hydrogen peroxide <8% solution used for mold remediation, as a bacteriacide, virus inactivation, a fungicide as well as a sporicide solution when used with the SteraMist™ system. This patented solution can be used in any industrial, commercial and institutional setting, including production operations in pharmaceutical manufacturing inclusive of clean rooms, laboratories, and animal research facilities; hotel rooms, offices, cruise ships and recreational facilities. Sold in 1 gal. bottles, 4 bottles per case. Minimum order is 4 bottles.
Catalog No. NC1442001 Supplier Accuflow Systems Supplier No. BIT400
The BIT-400 a ready-to-use solution, is to be used strictly in conjuction with SteraMist™ disinfection/decontamination system. It is a Hydrogen peroxide <8% solution used for mold remediation, as a bacteriacide, virus inactivation, a fungicide as well as a sporicide solution when used with the SteraMist™ system. This patented solution can be used in any industrial, commercial and institutional setting, including production operations in pharmaceutical manufacturing inclusive of clean rooms, laboratories, and animal research facilities; hotel rooms, offices, cruise ships and recreational facilities. Sold in 1 gal. bottles, 4 bottles per case. Minimum order is 4 bottles.