Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ SterEM™ Sabouraud Dextrose Agar with Lecithin and Tween 80
SDP

Catalog No. 23001779 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-779 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-779 Supplier Hardy Diagnostics™ Supplier No. W595

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Irradiated Sabouraud Dextrose (SabDex) Agar with Lecithin and Tween is recommended for the detection and enumeration of yeasts and fungi from environmental sources in a clean room setting

  • Standard size (15X100mm) passive settling plate for air monitoring of yeasts or fungi
  • Ideal for environmental monitoring in a clean room or clean bench setting
  • Plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level
  • Optically clear polystyrene for distortion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding

Specifications

Appearance Translucent
Color Light Amber
Additive Lecithin and Tween™ 80
Form Agar
Format Agar Plate
Product Type Agar Plate
Target Organism Class Yeast, Molds, Fungi
Fill Volume 34 mL
Incubation Conditions 20°C to 25°C
Incubation Time Variable
No. per Pack 10
Packaging Type Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Candida albicans, Aspergillus brasiliensis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.