Learn More
Hardy Diagnostics SterEM Sabouraud Dextrose Agar, (SabDex Agar), USP, Irradiated, Triple Bagged
Shop All Hardy Diagnostics Products

Description
- Irradiated Sabouraud Dextrose (SabDex) Agar is recommended for the detection and enumeration of yeasts and fungi from environmental sources in a clean room setting
- SterEM™ Sabouraud Dextrose Agar (SabDex Agar), USP is a standard size (15 x 100 mm) plate for air monitoring of fungi (yeast and mold)
- The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level
- Features and Benefits:
- Triple bagged: Transparent PET outer and middle bag for distortion-free viewing, breathable transparent inner bag
- Polystyrene Petri plates feature raised rings for stacking ability and prevent sliding
- Sterile by irradiation, validated for Sterility Assurance Level (SAL) of 10⁻⁶
- Certificate of Analysis is available on the Hardy Diagnostics website 24/7
- Manufactured in a licensed cGMP facility with a quality management system certified to ISO 13485
Specifications
Specifications
| Form | Agar |
| Manufacturing Origin | US with Globally Sourced Materials |
| Product Type | Agar Plate |
| Sterility | Sterile |
| Target Organism Class | Fungi, Molds, Yeast |
| Test Type | Media Testing, Microbial Detection, Growth Promotion Testing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.