Learn More
Electron Microscopy Sciences Stereo Microscope Boom Stand, Binocular, 6.5:1 Zoom, 10x to 65X magnification
SDP
Supplier:  Electron Microscopy Sciences GL6206
Encompass_Preferred
The remarkably clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design. The GL Series Greenough design produces lifelike images in a large field of view and presents upright or erect images with astounding depth perception. A long (100 mm) working distance facilitates observation.
Type: Binocular
Range: 6.5:1 Zoom
Magnification: 10x to 65X
Catalog No. 50-342-42