Learn More
Electron Microscopy Sciences Stereo Microscope Boom Stand, Binocular, 7:1 Zoom, 6.5x to 45X magnification
SDP
Catalog No. 5034255
Encompass_Preferred
The remarkably clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design. The GL Series Greenough design produces lifelike images in a large field of view and presents upright or erect images with astounding depth perception. A long (100 mm) working distance facilitates observation. Articulating Boom.
Type: Binocular
Range: 7:1 Zoom
Magnification: 6.5x to 45X
Catalog No. 50-342-55 Supplier Electron Microscopy Sciences Supplier No. GL7208
The remarkably clear, non-distorted 3D image resolved by the GL Series optics is a result of advanced technologies in optical design. The GL Series Greenough design produces lifelike images in a large field of view and presents upright or erect images with astounding depth perception. A long (100 mm) working distance facilitates observation. Articulating Boom.
Type: Binocular
Range: 7:1 Zoom
Magnification: 6.5x to 45X