Learn More
Med Vet International Stereo Microscope 1X/3X w/ Led Light
Catalog No. 502830195
Encompass_Preferred
Head can be raised or lowered on post to accommodate various sized specimens. Versatile illumination system offers incident (top) lighting or transmitted (bottom) lighting through the frosted glass stage plate. Precision optics provide a wide field of view with good depth and high resolution. Now offering two models with choices of magnification to fill any needs.1X & 3X objectives for 10X/30X magnification2X & 4X objectives for 20X/40X magnification
Catalog No. 50-283-0195 Supplier Med Vet International Supplier No. SMP13L
Head can be raised or lowered on post to accommodate various sized specimens. Versatile illumination system offers incident (top) lighting or transmitted (bottom) lighting through the frosted glass stage plate. Precision optics provide a wide field of view with good depth and high resolution. Now offering two models with choices of magnification to fill any needs.1X & 3X objectives for 10X/30X magnification2X & 4X objectives for 20X/40X magnification