Learn More
RWD LIFE SCIENCE INC STEREOTAXIC INSTRUMENT
SDP
Catalog No. NC3404297
Encompass_Preferred
RWD standard stereotaxic instrument with the classical U-type base design has been trusted in laboratory settings for over 22 years. The instrument offers a displacement accuracy of 100 um, and its versatile U-shaped frame allows for stereotaxic surgery on various small animal species by simply swapping adaptors and ear bars. The complete configuration includes the standard stereotaxic instrument, adaptor, ear bars, holder, and nosecone mask. Additionally, the single manipulator can be upgraded to a dual manipulator setup by adding a second manipulator. For enhanced precision, you can also contact us to upgrade to a digital display model with an accuracy of 10 um.
Catalog No. NC3404297 Supplier RWD LIFE SCIENCE INC Supplier No. 68801
RWD standard stereotaxic instrument with the classical U-type base design has been trusted in laboratory settings for over 22 years. The instrument offers a displacement accuracy of 100 um, and its versatile U-shaped frame allows for stereotaxic surgery on various small animal species by simply swapping adaptors and ear bars. The complete configuration includes the standard stereotaxic instrument, adaptor, ear bars, holder, and nosecone mask. Additionally, the single manipulator can be upgraded to a dual manipulator setup by adding a second manipulator. For enhanced precision, you can also contact us to upgrade to a digital display model with an accuracy of 10 um.