Learn More
World Precision Instrument WPI Standard Mouse Stereotaxic Instrument, Non-Rupture Ear Bars (Non-Digital)
SDP
Catalog No. 502539072
Encompass
The Stereotaxic Instrument, Non-Rupture Ear Bars, Mouse uses a more traditional low profile hold for the head and snout of the mouse using the traditionl 'U' Frame. Non-Rupture (45°) Ear Bars and a mouse specific bite bar for use on the traditional frame are included.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9072 Supplier World Precision Instrument Supplier No. 505388
The Stereotaxic Instrument, Non-Rupture Ear Bars, Mouse uses a more traditional low profile hold for the head and snout of the mouse using the traditionl 'U' Frame. Non-Rupture (45°) Ear Bars and a mouse specific bite bar for use on the traditional frame are included.
View our Stereotaxic Frame Compatibility Chart