Learn More
World Precision Instrument Steri-Lite Bead Sterilizer-ST5193-EU
SDP
Catalog No. 502538542
Encompass
WPI's Steri-Lite Glass Bead Sterilizer is ideal for the sterilization of small research instruments (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms.
Catalog No. 50-253-8542 Supplier World Precision Instrument Supplier No. ST5193EU
WPI's Steri-Lite Glass Bead Sterilizer is ideal for the sterilization of small research instruments (forceps, scissors, etc.). When the chamber is filled with the included glass beads, high temperatures (up to 300°C) can be used to eliminate the presence of bacteria, spores and other microorganisms.