Learn More
Andwin Scientific STERIGAGE STM INTEGRATOR 500PK
SDP
Catalog No. NC9747981
Encompass_Preferred
The 3M Comply Sterigage is a chemical indicator for internal or external use in steam sterilization cycles. This product features a moving front design with easily interpretable Accept/ Reject result. It also complies with the USP requirement for monitoring steam parametric release processes. This is sold in a pack of 500.
Catalog No. NC9747981 Supplier Andwin Scientific Supplier No. 34010019
The 3M Comply Sterigage is a chemical indicator for internal or external use in steam sterilization cycles. This product features a moving front design with easily interpretable Accept/ Reject result. It also complies with the USP requirement for monitoring steam parametric release processes. This is sold in a pack of 500.