Learn More
Enzo Life Sciences Sterigmatocystin (1mg). CAS: 10048-13-2
SDP
Supplier:  Enzo Life Sciences ALX630116M001
Encompass_Preferred
Mycotoxin produced by strains of the common molds. Inhibitor of DNA synthesis. Induces sister chromatid exchanges in bone marrow cells of mice. Causes necrosis. Alternative name: 3a 12c-Dihydro-8-hydroxy-6-methoxy-7H-furol 3' 2' 4 5 furo 2 3-c xanthen-7-one. Purity: ≥98% (TLC, HPLC). Appearance: Pale yellow solid. Solubility: Soluble in DMSO (10mg/ml), chloroform (71mg/ml), pyridine (18mg/ml) or acetone (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9643