Learn More
AdipoGen Sterigmatocystin
SDP
Supplier:  AdipoGen BVT0171M001
Encompass_Preferred
Chemical. CAS 10048-13-2. Formula C18H12O6. MW 324.3. Isolated from Aspergillus sp. strain WDMH51. Intermediate of the biosynthetic pathway to aflatoxin B1. Mycotoxin. DNA synthesis inhibitor. Anticancer compound. Cytotoxic, carcinogenic and mutagenic. Acyl-CoAcholesterol acyltransferase 2 ACAT2 inhibitor. Shown to induce apoptosis in human peripheral lymphocytes and necrosis in rat liver. Induces sister chromatid exchanges in murine bone marrow cells.
Catalog No. 50-256-2078