Learn More
Molecular Devices Molecular Devices IMAP TR-FRET Kit, Screening Express, Progressive Binding
Catalog No. 5000014442
Encompass_Preferred
The Molecular Devices IMAP TR-FRET Screening Express Kit (R8160) is an antibody-free, homogeneous TR-FRET assay kit that combines IMAP progressive binding chemistry with time-resolved FRET detection to measure kinase, phosphatase, and phosphodiesterase activity in a mix-and-read workflow. The kit is optimized for high-throughput screening and supports multiple microplate formats.
  • Antibody-free TR-FRET assay format for phosphorylation detection.
  • Kit capacity of 8,000 data points.
  • Includes progressive binding reagent, TR-FRET donor, and concentrated buffers.
  • Optimized for white 384-well microplates; adaptable to 96- and 1536-well plates.
  • Store components at 4°C; do not freeze the binding reagent.
  • Intended for research use only.
Catalog No. 50-000-14442 Supplier Molecular Devices Supplier No. R8160
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices IMAP TR-FRET Screening Express Kit (R8160) is an antibody-free, homogeneous TR-FRET assay kit that combines IMAP progressive binding chemistry with time-resolved FRET detection to measure kinase, phosphatase, and phosphodiesterase activity in a mix-and-read workflow. The kit is optimized for high-throughput screening and supports multiple microplate formats.
  • Antibody-free TR-FRET assay format for phosphorylation detection.
  • Kit capacity of 8,000 data points.
  • Includes progressive binding reagent, TR-FRET donor, and concentrated buffers.
  • Optimized for white 384-well microplates; adaptable to 96- and 1536-well plates.
  • Store components at 4°C; do not freeze the binding reagent.
  • Intended for research use only.