Learn More
BioSpec Products STERILE, 2ML VIAL, 100/PK
SDP
Catalog No. NC0557839
Encompass_Preferred
Our 2.0 ml, sterile screw cap with o-ring seal, microcentrifuge vials are made of inert polypropylene and have straight walls with a sharp conical bottom.
Catalog No. NC0557839 Supplier BioSpec Products Supplier No. 10831
Our 2.0 ml, sterile screw cap with o-ring seal, microcentrifuge vials are made of inert polypropylene and have straight walls with a sharp conical bottom.